3M Solutions - Respiratory
Respiratory
Science driven innovation helping to provide comfort, fit and protection
3M offers you the widest range of disposable, reusable, and powered & supplied air respirators for the widest range of industrial and other applications, with unequalled pedigree, gained in the toughest conditions.
3M™ Secure Click™ Reusable Half Mask HF-800 Series
Designed with smart and intuitive features 3M Secure Click Half Mask Reusable Respirator HF-800 Series is simple, comfortable and reliable. Available in three sizes: small, medium, and large, our respirators all come with an optional speaking diaphragm. The new 3M Secure Click Series features a quick, simple cartridge/filter connection. It works just like a seat belt. Simply align the connections and push until you hear a click and feel confident that filters and cartridges are installed properly.
Resources
3M™ Versaflo™ Intrinsically Safe Powered Air Respirator TR-800+ Series
Use 3M Versaflo Intrinsically Safe Powered Air Respirator TR-800 Series for protection when working in hazardous atmospheres. As part of the Versaflo family, when combined with Versaflo headtops, belts, and other accessories our air turbo becomes part of a CE-approved respirator system featuring; multiple speeds, audible and visual alarms, and the ability to work with a wide variety of gas, vapour and particulate filters. Comfort is maximised with three user selectable flow rates and controlled airflow, and it’s easy to identify and differentiate from non-IS versions of the Versaflo product family thanks to bright yellow touchpoints. The TR-800 is approved to Zone 0 requirements and so may be suitable for areas where potentially explosive atmospheres are expected and permanent in normal operations.
See the product
Resources
3M™ Versaflo™ Powered Air Respirator TR-600 Series
The 3M™ Versaflo™ Powered Air Turbo TR-600 takes the entire system to new levels of confidence, control and comfort. Representing an advanced evolution of technology with increased excellence in powered air respiratory safety, your world of protection just expanded and is better than ever. Depending on the type of filter used it provides protection against particles, gases or vapours, or against a combination of these.
See the product
Resources
  • Read the Brochure
  • Read the Specification
  • Watch the video
3M™ Versaflo™ S-Series Hoods & Headcovers
3M™ Versaflo™ S-Series Hoods and Headcovers feature adjustable suspensions (most models) and soft comfort pads help provide excellent fit, stability and tracking. Suspension hoods and headcovers include wide faceseals with soft elastic for improved comfort and reduced air flow noise. Reusable suspension allows wearers to modify airflow distribution inside the hood to suit personal comfort needs.
More