Viscosity standards
New range of certified, accurate and traceable viscosity standards. These products can be used for calibration, control, verification, qualification or method validation of kinematic and dynamic viscosity measurement instruments (both manual and automatic).
  • Manufactured and certified according to ASTM D2162 using Ubbelohde Master viscometers
  • Each standard is certified for kinematic viscosity (mm2/s,cSt), dynamic viscosity (cP) and density (g/ml) at a range of temperatures
  • Attractive 500 ml secure packaging
  • Manufactured from high quality, stable base oils and additives
  • Certificates of Analysis and Safety Data Sheets available online for every batch  manufactured
    Description Pk Cat. No. Promo price Quantity
    Viscosity standards (cSt) reads; 0,47 20 °C, 0,45 25 °C, 0,41 37,78 °C, 0,4 40 °C 500 ml 85067.260E  
    Viscosity standards (cSt) reads; 0,74 20 °C, 0,7 25 °C, 0,61 37,78 °C, 0,6 40 °C 500 ml 85068.260E  
    Viscosity standards (cSt) reads; 1,3 20 °C, 1,2 25 °C, 1 37,78 °C, 0,97 40 °C, 0,87 50 °C 500 ml 85069.260E  
    Viscosity standards (cSt) reads; 20 20 °C, 16 25 °C, 11 37,78 °C, 10 40 °C, 7,5 50 °C 500 ml 85070.260E  
    Viscosity standards (cSt) reads; 320 20 °C, 230 25 °C, 110 37,78 °C, 98 40 °C, 61 50 °C 500 ml 85071.260E  
    Viscosity standards (cSt) reads; 3300 20 °C, 2300 25 °C, 1100 37,78 °C, 940 40 °C, 560 50 °C 500 ml 85072.260E  
    Viscosity standards (cSt) reads; 58 000 20 °C, 36 000 25 °C, 10 000 40 °C, 4900 50 °C 500 ml 85073.260E  
    Viscosity standards (cSt) reads; 30 20 °C, 24 25 °C, 15 37,78 °C, 14 40 °C, 10 50 °C 500 ml 85074.260E  
    Viscosity standards (cSt) reads; 400 20 °C, 300 25 °C, 160 37,78 °C, 140 40 °C, 90 50 °C 500 ml 85075.260E  
    Viscosity standards (cSt) reads; 4900 20 °C, 3500 25 °C, 1600 37,78 °C, 1400 40 °C, 30 50 °C 500 ml 85076.260E  
    Viscosity standards (cSt) reads; 100 000 20 °C, 64 000 25 °C, 18 000 40 °C, 8500 50 °C 500 ml 85077.260E  
    Viscosity standards (cSt) reads; 2,9 20 °C, 2,6 25 °C, 2,1 37,78 °C, 2 40 °C, 1,7 50 °C 500 ml 85078.260E  
    Viscosity standards (cSt) reads; 790 20 °C, 580 25 °C, 280 37,78 °C, 250 40 °C, 160 50 °C 500 ml 85079.260E  
    Viscosity standards (cSt) reads; 7900 20 °C, 5800 25 °C, 2800 37,78 °C, 2500 40 °C, 1500 50 °C 500 ml 85080.260E  
    Viscosity standards (cSt) reads; 59 20 °C, 47 25 °C, 27 37,78 °C, 25 40 °C, 18 50 °C 500 ml 85081.260E  
    Viscosity standards (cSt) reads; 88 20 °C, 66 25 °C, 35 37,78 °C, 32 40 °C, 21 50 °C 500 ml 85082.260E  
    Viscosity standards (cSt) reads; 980 20 °C et 710 25 °C, 340 37,78 °C, 310 40 °C, 190 50 °C 500 ml 85083.260E  
    Viscosity standards (cSt) reads; 6,7 20 °C, 5,8 25 °C, 4,2 37,78 °C, 4 40 °C, 3,2 50 °C 500 ml 85084.260E  
    Viscosity standards (cSt) reads; 19 000 20 °C, 12 000 25 °C, 4000 37,78 °C, 3400 40 °C, 1700 50 °C 500 ml 85085.260E  
    Viscosity standards (cSt) reads; 1400 20 °C, 1000 25 °C, 470 37,78 °C, 410 40 °C, 250 50 °C 500 ml 85086.260E  
    Viscosity standards (cSt) reads; 110 20 °C, 87 25 °C, 48 37,78 °C, 44 40 °C, 30 50 °C 500 ml 85087.260E  
    Viscosity standards (cSt) reads; 28 000 20 °C, 17 000 25 °C, 6000 37,78 °C, 5100 40 °C, 2500 50 °C 500 ml 85088.260E  
    Viscosity standards (cSt) reads; 14 20 °C, 12 25 °C, 8 37,78 °C, 7,5 40 °C, 5,8 50 °C 500 ml 85089.260E  
    Viscosity standards (cSt) reads; 210 20 °C, 160 25 °C, 83 37,78 °C, 75 40 °C, 50 50 °C 500 ml 85090.260E  
    Viscosity standards (cSt) reads; 2600 20 °C, 1800 25 °C, 850 37,78 °C, 750 40 °C, 450 50 °C 500 ml 85091.260E  
    Viscosity standards (cSt) reads; 43 20 °C, 34 25 °C, 20 37,78 °C, 18 40 °C, 13 50 °C 500 ml 85092.260E  
    Viscosity standards (cSt) reads; 550 20 °C, 400 25 °C, 200 37,78 °C, 180 40 °C, 110 50 °C 500 ml 85093.260E  
    Viscosity standards (cSt) reads; 8400 20 °C, 5300 25 °C, 1900 37,78 °C, 1600 40 °C, 810 50 °C 500 ml 85094.260E  
    Viscosity standards (cSt) reads; 4,4 20 °C, 3,9 25 °C, 3 37,78 °C, 2,9 40 °C, 2,4 50 °C 500 ml 85095.260E  
    Viscosity standards (cSt) reads; 10 20 °C, 8,7 25 °C, 6 37,78 °C, 5,7 40 °C, 4,4 50 °C 500 ml 85097.260E  
    Viscosity standards (cSt) reads; 160 20 °C, 120 25 °C, 60 37,78 °C, 54 40 °C, 35 50 °C 500 ml 85098.260E  
    Viscosity standards (cSt) reads; 1800 20 °C, 1300 25 °C, 590 37,78 °C, 520 40 °C, 310 50 °C 500 ml 85099.260E  
    Viscosity standards (cSt) reads; 41 000 20 °C, 25 000 25 °C, 8000 37,78 °C, 6700 40 °C, 3200 50 °C 500 ml 85100.260E