Balance Calibration
Brand-independant calibration service for balances and weighing equipment
Balances and scales should be properly installed and calibrated before daily operation. To ensure the accuracy and for a longer lifetime of the balance we recommend an annual preventive maintenance and calibration contract. VWR can provide you with a complete range services for balances from all major manufacturers.
VWR Compliance & Metrology services can ensure:
• ISO 17025 accredited and traceable calibrations
• Accurate and reliable results
• Qualified procedures and technicians
• Calibration certificates with individual results for linearity, excentral weighing, repeatibility and measurement uncertainty
If you want to perform the calibration of your balances and scales yourself we can provide you with certified external weights. The weights are provided with an ISO17025 calibration or traceable certificate.VWR has a complete offering of certified weights.