Disposable Overalls
Disposable overalls protect sensitive products as well as the operators. They are manufactured specifically to prevent particle contamination and static build up. 
Disposacle cleanroom overalls are made from different materials (Tyvek, Polypropylene, breathable SMS Fabric, Polyethylene, Nonwoven, Clean Tough Fabric).
Sterile and non sterile versions of disposable cleanroom overalls with elasticated wrists and ankles are available.
Upon request with integrated hood. Single hoods are available.
Product selection
Disposable Overalls
Please choose from a list of products and refine your search by attributes...