BOD incubators
All BOD methods require samples to be kept at 20 °C (68 °F) for 5 days. VWR offers a wide range of temperature-controlled incubators.
The Lovibond® range
6 models; 2 to 40 °C (0,1 °), 135 to 445 l
The Velp range
6 models; fixed or 3 to 50 °C (0,1 °), 120 to 215 l
The WTW range
  • OxiTop® Box: Fixed at 20 °C ±0,5 °C
  • 5 models; 10 to 40 °C (0.5 °), 180 to 500 l