So Much More Than a Clean Surface…
So Much More Than a Clean Surface…
Contec Inc is an ISO 9001:2015 registered manufacturer of contamination control products for critical cleaning and disinfection in controlled environments worldwide. Contec’s extensive cleanroom product range of presaturated and dry wipes, mops, disinfectants and swabs; both sterile and non sterile is ideal for the many cleaning and disinfecting tasks in critical environments.
Contec cleanroom products are used in a variety of industries including pharmaceutical, medical device, biopharma, microelectronics, optics, semiconductor and electronics.
Alcohols, Disinfectants and Detergents
A wide range of disinfectants and solutions for critical and sterile environments. From hydrogen peroxide solutions and IPA, to fast-acting validated sporicidal disinfectants. Contec can also offer a sterile neutral detergent solution.
Cleanroom Wipes & Swabs
Sterile and non-sterile, dry and presaturated, knitted, nonwoven wipes and microfibre wipes, as well as a range of swabs of different lengths, head sizes, and materials.
Cleanroom Mopping Systems
A broad range of wall and ceiling mops, buckets, trolleys and tools for critical and cleanroom environments, whether disinfecting or cleaning floors, walls, ceilings, or inside bio-safety cabinets and isolators.
Our Featured Products
Contec ReFIBE™ 100% recycled polyester wipes
The 1st sustainable cleanroom polyester wipe made from recycled post-consumer plastic bottles. ReFIBE™ are 100% knitted polyester wipes with sealed edges, with each case stopping up to 840 bottles from going to landfill.
BPR authorised 70% IPA presaturated wipes
Presaturated wipes can offer many benefits over bottled alcohol and dry wipes. They reduce solvent use and increase convenience in hand wiping, whilst providing exceptional solvent control. Contec's 70% IPA presaturated wipes are all authorised to the EU
Contec ProChlor Sporicidal Disinfectant
Contec ProChlor is a revolutionary sporicide achieving a 100% kill against spores in under 1 min. A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts, viruses
Contec EasyReach™ Mini Environment Cleaning Tools
Designed for cleaning isolators, laminar air flow hoods, glove boxes and other mini environments, Contec’s lightweight, stainless steel, EasyReach™ Cleaning Tool is easy to use and improves access to hard to reach areas.
Videos
Literature
Webinars
Whitepapers, Articles