Turbidity
A simple and undeniable indicator of water quality change
The measurement of turbidity is a key test of water quality: drinking water – waste water monitoring – industrial waters and more…
Reliable turbidity monitoring is an essential component of drinking water production, and public health and safety organizations around the world recognize the importance of measuring this parameter.
A sudden change in turbidity may indicate an additional pollution source (biological, organic or inorganic) or may signal a problem in the water treatment process.
Are you having questions on the topic Turbidity or are you a beginner and need to know more?  Click below to download the "Theory of turbidity" booklet.
In practical applications significant problems can introduce interference and errors that reduce the accuracy of any instrument. To ensure the instrument is operating properly and providing the most accurate answers possible, it is important to verify its calibration.  Click below to download the "Technical information on standards"  booklet.
Literature
Comparison between TU5 and TL23 Laboratory Turbidimeters
- Product Details
TU52 bench top turbidimeter, Hach
The most precise and advanced turbidimeter on the market
  • Patented 360°x90° optics
  • The most sensitive and the fastest
  • The benchmark for clean water
- Product Details
The Benchtop Turbidimeter TL23 series, Hach
Trusted measurement for wide range of turbidity applications
A turbidimeter that simplifies testing in the most demanding waste water and industrial applications
  • Easy to use intuitive full color touch screen
  • Easy data management and export capabilities
  • Easy to be right
Delivered with sample cells, standards, accessories
- Product Details
2100Q portable turbidimeter, Hach
Ideal for use in the field or around the plant
  • Easy to carry around
  • Takes multiple measurements from various collection points in your treatment process
  • Intuitive user interface: Easy to measure, calibrate and verify
  • Easy storing and transferring of data
- Product Details
The new portable VWR TIR200
For fast, precise, easy On-Site testing
From drinking water to waste water, to be used for both colored and colorless liquids.  
Conforms to EN ISO 7027.
- Product Details
WTW, IDS turbidity sensors – digital & wireless ready
VisoTurb® 900-P: Digital IDS turbidity measurement with a practical turbidity sensor
  • Convenient turbidity sensor to measure turbidity in surface waters and process
  • Simple 2 or 3 point calibration
  • Suitable for WTW MultiLine® IDS multi-parameter measurements on site
Full range of turbidity meters
Full range of turbidity standards