Greener solutions for Controlled Environments
The cleanroom is inherently a challenging environment to be environmentally conscious. We use a lot of energy and rely on single-use items and additional packaging to reduce the risk of contamination events. It is also difficult to make changes when articles are written into SOP's or would require re-validation effort to change. There are, however, a number of new innovations coming to the market now where the environmental impact is considered and every step towards reducing our carbon footprint. Whether that be through using less or buying better, it is a step in the right direction.
Cleanroom wipes
Contec Refibe
Non sterile and sterile 100% knitted, laundered polyester cleanroom wipes made from 100% recycled consumer plastic water bottles. Each pack of wipes stops 35+ bottles from going to landfill
Texwipe Revolve
Dry or pre-wetted wipes, mop covers and pads, available sterile and non-sterile. Made from post-consumer polyester (rPET) yarn, made by upcycling plastic water bottles
Diversey Oxivir wipes
The active ingredient, hydrogen peroxide, breaks down into environmentally-friendly water and O2. The wipes and tubs are recyclable.
Cleanroom stationary and packaging
VWR PureImage cleanroom paper
Recyclable as there is no latex coating and comes with more sustainable
EcoBond cleanroom paper
A recyclable version of BCR® cleanroom bond paper. EcoBond® is made from cellulose and is recommended for use in ISO Class 3 to 7
Flooring and matting
VWR Purestep mats EnviroTack
Contains GMO free, renewable plant based material that reduces CO2 emissions Made with 20% specially designed biohybrid resin. Adhesive mats are also recyclable after use
Intelligent sole cleaning machine
Eliminates the need for tacky mats or shoe covers, so less disposable waste Simple to install, easy to clean, high particle removal
Micronova Novaclean detergent
Recyclable as well as safer for human and environmental health