Microbiology New Products
VWR® MPP15: Simplifying Culture Media Preparation
The VWR Medium Preparator Premium 15 (MPP15) is equipped with everything that simplifies and speeds up the production of culture media. The medium is effectively mixed, heated, sterilised and quickly cooled to the desired filling temperature. For the filling process, the medium is kept stable at the selected temperature. The MPP15 can also be used to for autoclaving, sterilisation of small objects and liquids in separate containers, or as a heating bath.
Additional Information
  • Product page
  • Brochure
VWR® Automatic Colony Counters
Why would you use an automatic colony counter?
Counting colonies automatically gives you consistency, accuracy, speed and efficiency.
Our new range of automatic colony counters includes units from low cost for rapid count of pour plates, spread plates and spiral plates, up to high-end, extended applications and IQ/OQ. 
Additional Information
  • Product page
  • Brochure
  • Reasons to use
T-Lab Eco: The Economic Laboratory Autoclave
Tuttnauer developed the T-Lab Eco especially for educational institutions and research facilities, with simplicity and economy in mind.
This vertical floor-standing autoclave covers the fundamental needs for general lab sterilisation with the aim of increasing the productivity of your laboratory, combining the best universal capacity size with the optimisation of resources such as water, power and operating time results. 
Additional Information
  • Brochure
Improve Your Enumeration Assays With The CytoFLEX Flow Cytometer
The CytoFLEX platform's advanced optics and fluidics enables the instrument to resolve small particles down to 200 nm, and count bacteria as small as 0.5 µm without the need for counting beads. Additionally, information provided on viability and vitality of the cells allow manufacturers to save time and money.
Portable MinION™ Sequencing Devices
Rapidly identify and fully characterise microorganisms from culture or complex metagenomic samples using affordable, portable MinION sequencing devices.
Real-Time analysis for rapid access to actionable results. 
Additional Information
VWR® Lockable Contact Plates (Screw-On Lid)
For pharmaceutical industries, with triple-bagged sterile packaging and lockable lid, ideal for environmental control of air, surfaces, gloves and overalls inside restricted areas like isolators and cleanrooms. Packed in bags of 5 plates.
Additional Information