Karl Fischer titration for water determination
Choice of compact volumetric or coulometric Karl Fisher titrators and related reagents for water determination.
  • Volumetric Karl Fischer titrators are for medium to large volumes of water (mg range) combining fast and precise water content determination with easy operation
  • Coulometric Karl Fischer titrators are for small water volumes (µg range) combine fast and precise water content determination with very simple operation
  • Karl Fischer reagents
Volumetric Karl Fischer Titrators
Routine Karl Fischer titrators combine fast and precise water content determination with easy operation...
Karl Fischer reagents and titrants
A wide range of volumetric and coulometric Karl Fischer reagents
Karl Fischer standards
A wide range of Karl Fischer standards for volumetric and coulometric titration