Single-Use Samplers For The Highest Standards In Sampling
Cleaning of samplers is a demanding, time-intensive task, especially when the material is greasy, strongly adhesive or toxic. Single-use is the solution
Sampling lances - for liquids
Single-use samplers for thin liquids to viscous fluids.
DispoDippers and DispoLadles
Dippers combine sampler and sampling container in one device, while ladles have embossed scales and two spouts for right- or left-handed users.
Bürkle Bio line - The purity of disposable products and ecological sustainability combined perfectly..
Made from renewable and 100% recyclable raw materials these products help to combine highest quality demands on sampling tools and sustainability.
Disposable forceps for precise lab work
Ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.