UV/VIS spectroscopy
UV/Vis analysis with a spectrophotometer can require different types of chemicals. First, a specific grade of solvents is developed to avoid optical impurities of your sample. Second, the spectrophotometer needs to be calibrated using UV/Vis standard kits. Third, color reference solution kits can save you time in preparing the reference solutions yourself.
Product Categories
Solvents for UV/Vis spectroscopy
High purity solvents specially designed for spectroscopy applications that demand the highest spectral purity.
UV/VIS Standards
Be sure of your results with our complete range to calibrate spectrophotometers on the control of absorbance, resolution power, stray light and wavelength accuracy
Color standard solutions
In petrochemistry mainly, we can offer complete range of "ready to use" solutions for use as calibration and/or quality control standards according to ASTM, Saybolt, Platinum-coblat and European Pharmacopoeia (Ph Eur)
Color standard in kits
These solutions are developed and produced according to chapter 2.2.2. "Degree of coloration of liquids" of the European Pharmacopeia.
FAQ
Different standards in ampoules delivered in kits with a complete Certificate of Analysis are available to check
parameters such as:
  • Absorbance control
  • Stray light testing
  • Resolution power testing
  • Wavelength testing
Literature
Additional Resource