Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carboys, narrow neck, round, with screw caps

Supplier: VWR Collection

LDPE, HDPE or PP, translucent with purple screw caps. Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder handle. Graduated to identify the volume dispensed.

LDPE carboys
Preferred for high purity applications as it has high resistance to chemicals (widely used in ophthalmic product range, like eye droppers, nasal spray).
Preferred resin for cold storage (up to −80 °C), working temperature +80 to −80 °C
Resistance to physical impacts (products will not break even if it falls down by accidentally, where as PP carboys may lead to crack at negative temperature)
LDPE product will bounce back to its shape as its high flexible material
Widely used in pharma industries

HDPE carboys
The high-density polyethylene (HDPE), disposable carboys are ideal for storing and transfer of sterile fluids and pharmaceutical/biotech reagents.
Good for cold storage (up to −80 °C), working temperature of +120 to −80 °C
High impact strength even at negative temperature
Widely used for long storage due to high chemical resistance property
Widely used in food & milk industries

PP carboys
Ideal for sterile water storage, polypropylene (PP) carboys can be autoclaved before refilling to stop recurring bacterial growth
Excellent chemical resistance
Working temperature 0 to 135 °C
Autoclavable

Note: Polypropylene carboys are autoclavable. LDPE and HDPE carboys are not autoclavable. Always disassemble cap from carboy before autoclaving.

Resin meets US FDA 21 CFR, U SP Class VI




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now

LDPE carboys
Capacity Type Neck I-Ø Thread Pk VWR Catalog Number Quantity
10 l non sterile 65 mm 83 mm 1 216-1704
15 l non sterile 65 mm 83 mm 1 216-1918
20 l non sterile 65 mm 83 mm 1 216-1706
50 l non sterile 65 mm 83 mm 1 216-1738
25 l non sterile 65 mm 83 mm 1 216-1908

HDPE carboys
Capacity Type Neck I-Ø Thread Pk VWR Catalog Number Quantity
10 l non sterile 65 mm 83 mm 1 216-1736
20 l non sterile 65 mm 83 mm 1 216-1737
20 l sterile 65 mm 83 mm 1 216-1729

PP carboys
Capacity Type Neck I-Ø Thread Pk VWR Catalog Number Quantity
10 l non sterile 65 mm 83 mm 1 216-1703
20 l non sterile 65 mm 83 mm 1 216-1701
50 l non sterile 65 mm 83 mm 1 216-1702
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty