Would you like to log in again?
Microplate centrifuge, PCR Plate Spinner
Supplier: VWR Collection
The PCR Plate Spinner offers a personal sized centrifuge designed to spin down PCR plates.
  • Space saving design; quiet, simple operation
  • Easy plate loading system
  • Quickly spins down droplets and condensation
  • Use before and after thermal cycling to improve PCR yield
  • Cold room compatible
This unique centrifuge is equipped with a dual position vertical rotor that accepts a wide variety of PCR plates; 96-well, 384-well, skirted and non skirted plates are compatible with the Plate Spinner. The rotor features integral plate guides, ensuring that the plates stay vertical and do not tip inside the rotor.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description Pk VWR Catalog Number Quantity
PCR Plate Spinner, EU-plug 1 521-1648
PCR Plate Spinner, UK-plug 1 521-1649
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Model PCR Plate Spinner
Max. capacity 2 PCR plates
Max. speed (min-1) 2500
Max. RCF (×g) 500
Power supply 230 V / 50-60 Hz
W×D×H (mm) 191×208×183
Weight (kg) 3
ACCESSORIES
Expand All / Collapse All
Accessories
Description Pk VWR Catalog Number Quantity
Adapter for strips, tubes or half plates 2 521-1650
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty