Would you like to log in again?
Pipette controller, Accurpette
Supplier: VWR Collection
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
  • Lightweight UV resistant body (180 g) and nose cones
  • Fills a 25 ml pipette in under three seconds on its fastest setting
  • Autoclavable silicone pipette holder
  • Replaceable hydrophobic membrane filter protects the unit against liquid influx and protects samples against contamination
  • Environmentally friendly rechargeable NiMH battery allow continuous usage for four hours; low battery light; rechargeable during use
The Accurpette pipette controller delivers efficient performance with a powerful but quiet motor to speed up large volume pipetting. The mode selection switch enables selection of high or low modes both allowing variable aspirate and dispense speeds (with blow out). In any mode setting the speed of suction and dispensing is controlled through the concave finger triggers, designed to provide a comfortable and positive grip and to require minimum effort. An additional gravity dispense mode is designed for use with ‘To Deliver’ (TD) pipettes. The Accurpette is now supplied with additional coloured nose cones to allow laboratory, application or user colour coding to minimise the risk of cross-contamination.
Ordering information: Supplied with charger, two spare hydrophobic filters (1× 0,45 µm, 1× 0,2 µm), bench stand/wall bracket and a coloured nose cone set.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description Pk VWR Catalog Number Quantity
Pipette controller Accurpette, EU-charger 1 612-4552
Pipette controller Accurpette, UK-charger 1 612-4553
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Pk VWR Catalog Number Quantity
Pack of three coloured nose cones (red, blue and pink) 1 612-4555
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty