Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Filter tips

Supplier: VWR Collection

All tips are made of high quality 100% pure virgin medical grade PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard. Sterile products are tested for endotoxin (pyrogen) contamination and certified to USP results. Resins are pre-tested for metal contamination. Certificates are on file by lot number for all test results.

  • A wider seal ensures that tips will seal easily and consistently without leaking, on old as well as new pipettes
  • Guaranteed free of detectable DNA, DNase/RNase, endotoxins and heavy metals



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now

Bulk
Tip style Volume Sterile Length Compatibility Packed Pk VWR Catalog Number Quantity
UltraFine™, extended 0,1 - 10 µl - 38,10 mm AF1 Bulk 1.000 732-0565
UltraFine™, graduated 0,1 - 10 µl - 31,42 mm AF1 Bulk 1.000 732-0561
Bevel Point™, graduated 0,5 - 10 µl - 44,83 mm AF2 Bulk 1.000 732-0517
Bevel Point™ 1 - 40 µl - 49,66 mm AF3 Bulk 1.000 732-0526
Bevel Point™ 1 - 50 µl - 49,66 mm AF4 Bulk 1.000 732-0567
Bevel Point™ 1 - 100 µl - 49,66 mm AF5 Bulk 1.000 732-0521
Bevel Point™ 1 - 160 µl - 49,66 mm AF6 Bulk 1.000 732-0571
Bevel Point™, wide orifice 1 - 200 µl - 49,66 mm AF7 Bulk 1.000 732-0558
UltraFine™ 1 - 200 µl - 52,07 mm AF7 Bulk 1.000 732-0539
Bevel Point™ 1 - 300 µl - 52,07 mm AF8 Bulk 1.000 732-0535
UltraFine™ 100 - 1000 µl - 88,90 mm AF9 Bulk 1.000 732-0532

Racked
Tip style Volume Sterile Length Compatibility Packed Pk VWR Catalog Number Quantity
UltraFine™, extended 0,1 - 10 µl - 38,10 mm AF1 10 racks, 96 each 960 732-0542
UltraFine™, graduated 0,1 - 10 µl - 31,42 mm AF1 12 racks, 96 each 1.152 732-0515
Bevel Point™, graduated 0,5 - 10 µl - 44,83 mm AF2 10 racks, 96 each 960 732-0518
Bevel Point™ 1 - 40 µl - 49,66 mm AF3 10 racks, 96 each 960 732-0527
Bevel Point™ 1 - 50 µl - 49,66 mm AF4 10 racks, 96 each 960 732-0568
Bevel Point™ 1 - 100 µl - 49,66 mm AF5 10 racks, 96 each 960 732-0522
Bevel Point™ 1 - 160 µl - 49,66 mm AF6 10 racks, 96 each 960 732-0529
Bevel Point™, wide orifice 1 - 200 µl - 49,66 mm AF7 10 racks, 96 each 960 732-0559
UltraFine™ 1 - 200 µl - 52,07 mm AF7 10 racks, 96 each 960 732-0540
Bevel Point™ 1 - 300 µl - 52,07 mm AF8 10 racks, 96 each 960 732-0536
Rainin® LTS™ style, low retention 1 - 300 µl - 61 mm AF8 10 racks, 96 each 960 732-2896
UltraFine™ 100 - 1000 µl - 88,90 mm AF9 6 racks , 96 each 576 732-0533

Racked, sterile
Tip style Volume Sterile Length Compatibility Packed Pk VWR Catalog Number Quantity
UltraFine™, extended 0,1 - 10 µl + 38,10 mm AF1 10 racks, 96 each 960 732-0543
UltraFine™, graduated 0,1 - 10 µl + 31,42 mm AF1 12 racks, 96 each 1.152 732-0516
Rainin® LTS™ style 0,1 - 20 µl + 48,26 mm AF13 10 racks, 96 each 960 732-0799
Micro 0,1 - 30 µl + 38,10 mm AF1 10 racks, 96 each 960 732-1100
Bevel Point™, graduated 0,5 - 10 µl + 44,83 mm AF2 10 racks, 96 each 960 732-0519
UltraFine™, graduated 0,5 - 10 µl + 49,66 mm AF16 10 racks, 96 each 960 732-1101
Bevel Point™ 1 - 40 µl + 49,66 mm AF3 10 racks, 96 each 960 732-0528
Bevel Point™ 1 - 40 µl + 49,66 mm AF3 5 racks, 192 each 960 732-0525
Bevel Point™ 1 - 50 µl + 49,66 mm AF4 10 racks, 96 each 960 732-0520
UltraFine™, FlexTop™, graduated 1 - 50 µl + 49,66 mm AF4 10 racks, 96 each 960 732-1105
Standard 1 - 100 µl + 49,66 mm AF5 10 racks, 96 each 960 732-1103
Bevel Point™, graduated 1 - 100 µl + 49,66 mm AF5 10 racks, 96 each 960 732-1102
UltraFine™, extended 1 - 100 µl + 83,82 mm AF5 6 racks, 96 each 576 732-1107
Bevel Point™ 1 - 100 µl + 49,66 mm AF5 10 racks, 96 each 960 732-0523
Bevel Point™ 1 - 160 µl + 49,66 mm AF6 5 racks, 192 each 960 732-0573
Bevel Point™ 1 - 160 µl + 49,66 mm AF6 10 racks, 96 each 960 732-0530
Rainin® LTS™ style 1 - 200 µl + 51,31 mm AF11 10 racks, 96 each 960 732-0800
UltraFine™ 1 - 200 µl + 52,07 mm AF7 10 racks, 96 each 960 732-0541
UltraFine™, extended 1 - 200 µl + 83,82 mm AF7 6 racks , 96 each 576 732-0610
Bevel Point™, wide orifice 1 - 200 µl + 49,66 mm AF7 10 racks, 96 each 960 732-0544
Bevel Point™ 1 - 300 µl + 52,07 mm AF8 10 racks, 96 each 960 732-0537
Rainin® LTS™ style, low retention 1 - 300 µl + 61 mm AF8 10 racks, 96 each 960 732-2898
UltraFine™, extended 1 - 300 µl + 71,12 mm AF8 6 racks , 96 each 576 732-0611
Rainin® LTS™ style 100 - 1000 µl + 88,90 mm AF12 6 racks, 96 each 576 732-0801
UltraFine™, FlexTop™ 100 - 1000 µl + 75,95 mm AF9 6 racks, 96 each 576 732-1106
UltraFine™ 100 - 1000 µl + 88,90 mm AF9 6 racks , 96 each 576 732-0534
Wide orifice 100 - 1000 µl + 88,90 mm AF9 6 racks, 96 each 576 613-0752
Aerosol-barrier 100 - 1200 µl + 88,90 mm K 6 racks, 96 each 576 613-0753
Sartorius® type 1250 µl + 88,90 mm - 6 racks, 96 each 576 732-2894

Pak Rak™ refill
Tip style Volume Sterile Compatibility Packed Pk VWR Catalog Number Quantity
UltraFine™, graduated 0,1 - 10 µl - AF1 Reload system 864 732-0563

Pak Rak™ Express refill
Tip style Volume Sterile Compatibility Packed Pk VWR Catalog Number Quantity
UltraFine™, graduated 0,1 - 10 µl - AF1 Pak Rak™ Express Refill 864 732-0564

Individually wrapped, sterile
Tip style Volume Sterile Length Compatibility Packed Pk VWR Catalog Number Quantity
UltraFine™, extended 0,1 - 10 µl + 38,10 mm AF1 Individually wrapped 200 732-0566
UltraFine™, graduated 0,1 - 10 µl + 31,42 mm AF1 Individually wrapped 200 732-0562
Bevel Point™ 1 - 40 µl + 49,66 mm AF3 Individually wrapped 200 732-0524
Bevel Point™ 1 - 50 µl + 49,66 mm AF4 Individually wrapped 200 732-0569
Bevel Point™ 1 - 100 µl + 49,66 mm AF5 Individually wrapped 200 732-0570
Bevel Point™ 1 - 160 µl + 49,66 mm AF6 Individually wrapped 200 732-0572
Bevel Point™, wide orifice 1 - 200 µl + 49,66 mm AF7 Individually wrapped 200 732-0560
UltraFine™ 1 - 200 µl + 52,07 mm AF7 Individually wrapped 200 732-0538
Bevel Point™ 1 - 300 µl + 52,07 mm AF8 Individually wrapped 200 732-0574
UltraFine™ 100 - 1000 µl + 88,90 mm AF9 Individually wrapped 200 732-0531
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty