2-Propanol, Prepsolv® for preparative chromatography, Supelco®

Supplier: Merck

Danger

Synonyms: IPA,Isopropyl Rubbing Alcohol,Isopropyl alcohol,Dimethyl carbinol,2-Hydroxy propane,Isopropanol,Propan-2-ol

M-Clarity™ quality level = MQ200

1.13350.2500
1.13350.2500EA 180 EUR
1.13350.2500
2-Propanol, Prepsolv® for preparative chromatography, Supelco®
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60,1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0,786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR