Verification standard for Spectroquant® Multy and Spectroquant® Move 100 colorimeters, Supelco®
Supplier: Merck
1.19302.0001
1.19302.0001EA 257 EUR
1.19302.0001
Verification standard for Spectroquant® Multy and Spectroquant® Move 100 colorimeters, Supelco®
Standards Colour Reference and Spectroscopy Standards
Designed to reassure the user about the accuracy and the reliability of Spectroquant® Multy and Spectroquant® Move 100 colorimeters.
Delivery information: Set includes seven sealed cells including a zero cell and stable coloured solutions for measuring absorption at 430, 530, 560, 580, 610 and 660 nm, certificate and instructions.