Dry heat indicator tapes
Supplier: KARTELL
1051
1051.EA 131 EUR
1051.
Dry heat indicator tapes
Sterilisation Indicators
Adhesive tape.
Specially designed for dry heat sterilisation (30 minutes at 155 °C). Colour change from white to brown.