Reusable earplugs, 1261/1271, Comets®
Supplier: Moldex
6421
111-1163EA 110 EUR
111-1163
Reusable earplugs, 1261/1271, Comets®
Earplugs Reusable Earplugs
These reusable earplugs provide protection against lower noise levels. They are designed to keep the user in touch with the environment.
  • Air bubble in the tip and soft material provide a comfortable seal
  • Plugs made of soft durable material – Washable and hygienic
  • Come with handy PocketPak storage container
SNR: 25 dB
Certifications: EN 352-2:2002
Caution: * One size fits all.