VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
331-0205EA 205 EUR
331-0205
VWR®, Bottles, Wide Neck, with Screw Cap
Bottles Plastic Bottles
PE, translucent with black PP caps.
  • Good chemical resistance
  • Good thermal resistance
  • Leakproof
Perfect for sample collection, storage and transport. Ideal for liquids, pastes and granular material. The extra wide mouth allows easy filling and cleaning.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.