Bunsen burner, Labogaz®
Supplier: GLASWARENFABRIK KARL HECHT
41396010
HECH41396010EA 120 EUR
HECH41396010
Bunsen burner, Labogaz®
Heaters Gas Burners
Bunsen burner with new assembling-system for more security to cartridges without safety valve, gas consumption 55 g/h.