Water purification systems, RiOs-DI® Clinical
Supplier: Merck
ZRDSVCL3BE ZRDSVCL3DK ZRDSVCL3FR ZRDSVCL3EU ZRDSVCL3GE ZRDSVCL3IT ZRDSVCL3SP ZRDSVCL3SZ ZRDSVCL3UK
MILLZRDSVCL3BEEA 0 EUR
MILLZRDSVCL3BE MILLZRDSVCL3DK MILLZRDSVCL3FR MILLZRDSVCL3EU MILLZRDSVCL3GE MILLZRDSVCL3IT MILLZRDSVCL3SP MILLZRDSVCL3SZ MILLZRDSVCL3UK
Water purification systems, RiOs-DI® Clinical
Water Purification Systems Pure and Ultra Pure Water Systems
This system is a very compact solution designed for low volume analysers. It efficiently combines reverse osmosis (RO) technology with deionisation (DI) resins to produce CLSI® Special Reagent Water. With high resistivity and low TOC (typically >10 MΩ.cm at 25 °C and <50 ppb before storage), this product water is of higher quality than either water produced by stills or service DI water.
  • Product water can be stored in the system’s integrated 6 L reservoir
  • Specifically designed to feed low-volume analysers using up to 20 L/day
  • Convenient installation anywhere in your laboratory with its compact design: wall-mounted or on the bench
  • Easily insert the pack in one movement with the patented locking mechanism
  • Backlit colour display provides just the information required
  • Provides an efficient alternative to buying and storing bottled water