Would you like to log in again?
Cleanroom Furniture
Space is at a premium in cleanrooms where every square metre is expensive. As a result there are no standard solutions and no standard sizes.
The stainless steel cleanroom furniture, which is electrically conductive, consists of vertical supports with solid faces and perforated horizontal tops to reduce disturbance of the vertically directed laminar flow. Moving and cleaning the furniture is also facilitated. The holes in the tops have a diameter of 8-10mm and are chamfered on both sides.
basan Service
We support you in the planning of your gowning rooms and the (re)construction of your personnel air-locks.