Would you like to log in again?
Safety cleanroom goggles
Gamma or EtO single use? Autoclavable or VHP reusable? Designed with your SOP and operators in mind, disposable and reusable, our goggles for cleanroom or controlled environment use are manufactured from strong, lightweight and comfortable cleanroom compatible materials in a choice of styles.
Safety Goggles
Both autoclavable and single use goggles have anti-fog systems, optically correct toughened lenses and indirect ventilation systems. They are suitable for those who wear glasses and are 100% latex-free. To ensure the highest quality, every pair of our sterile single use goggles is individually inspected and suitable for medical supplies, personal protection equipment or wherever safety and protective goggles are required.