Would you like to log in again?
Cleanroom Stationery
Due to a complete range of cleanroom compatible paper, notebooks and other writing materials, there need to be no restrictions on the recording and retaining of data and other information in cleanroom areas.
Stationery
Cleanroom notebooks and cleanroom sticky pads are made from cleanroom paper. The writing on both sides of the sheets possible. We remmonend our durable and resistant clipboards to faciliate the scribble of notes in your cleanrooms and controlled production areas.
Special cleanroom pens with a non-slip grip ease the writing even with cleanroom gloves. The ink (different colors are available) of the cleanroom pens is permanent, also on cold and wet surfaces (e.g. plastic bags, disposable labware) and quick drying. It will not smear but can be scrubbed off.