Would you like to log in again?
Disposable Cleanroom Garments
Protection of products and operators by disposable garments
Disposable garments are designed and manufactured from lightweight, low-linting and durable material, providing an excellent combination of both comfort and protection.
basan provides the full range of disposables for cleanrooms and controlled environments, covering the full head-to-toe range, including overalls, coats, hoods, sleeve protectors and boot covers. To suit all areas, the range is available in sterile and non-sterile.
Our recommendation:
Dispenser systems for the simple collection of cleanroom disposables such as hoods, facemasks, shoe covers or gloves.