Would you like to log in again?
Cleanroom Gloves and Fingercots
In cleanrooms, gloves and fingercots are used to protect sensitive products and also operators from contamination.
Gloves and Fingercots We offer a wide range of gloves for cleanrooms and controlled environments relating to technical and functional needs for medical supplies, personal protection or cleanroom. Performance requirements such as grip, comfort and length vary and influence glove selection for the different applications.
The sterile and non-sterile ISO Class 4 and 5 compatible gloves especially for life science cleanroom environments offer extreme comfort, strength and tactility. 
We strongly recommend to test the gloves in practical application before choosing a specific model.
Glove Liners
Use textile gloves as glove liners to reduce sweating while wearing sterile latex and nitrile cleanroom gloves.
Cuff Stripes
Cuff Stripes for an effective seal between glove and cleanroom garment.