Revco® ultra-low temperature freezers, Revco®, DxF series
Supplier: Thermo Scientific
471-1057EA 0 EUR
REVCDXF40040V
Revco® ultra-low temperature freezers, Revco®, DxF series
Revco® DxF upright −40°C ultra-low temperature freezers are ideally suited for life science, university, government and academic accounts where accessible controls and at-a-glance monitoring are preferred. A knee-level easy to use microprocessor control panel, monitoring system and optional chart recorder designed for price sensitive sample protection without compromising performance. These freezers offer easy four-button operation and a clear intuitive user interface. A simple temperature display and visual LED's, guide non-technical users through the set-point temperature and alarm parameter setting process.
  • Alarm indications includes: high and low temperature, power failure, door ajar, hot condenser, probe failure and low battery
  • Five interior compartment doors to reduce cold air loss, improve temperature recovery after door closings (five door option available)
  • Ergonomic, lockable door handle
  • Easy-to-remove air filter
Construction features include: 5 interior compartment doors which minimizes cold air loss/warm air intrusion during door openings, reinforced stainless steel interior and shelves, triple point door gasket, heavy-gauge, cold-rolled steel cabinets with powder coat paint finish, 5" foamed-in-place polyurethane insulation, ergonomically designed handle, convenient air filter location for inspection and changing, dual 2" lockable casters, surge suppression, automatic voltage compensator responds to high and low voltages, environmentally friendly HFC refrigerants, vacuum relief port on all upright freezers allows easy re-entry after door openings.
Optional backup systems inject liquid CO2 or LN2 when cabinet temperature warms to a preset level.
Temperature range: −10 to −40 °C