HPLC columns, Syncronis™ Phenyl

Supplier: Thermo Scientific
Syncronis™
HYPE97903-104630EA 836 EUR
HYPE97903-104630 HYPE97902-102130 HYPE97905-012101 HYPE97905-152130 HYPE97905-014001 HYPE97905-154630 HYPE97905-254630 HYPE97902-052130 HYPE97905-054630
HPLC columns, Syncronis™ Phenyl
Chromatography Columns
These columns provide alternative selectivity to C18 and are particularly useful for retention of aromatic compounds.

  • Double endcapped for extra surface coverage
  • Highly inert towards basic compounds

Surface area: 320 m²/g

Carbon load: 11%

pH range: 2 - 8
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR