Dearomatized hydrocarbons EXXSOL DSP 100/140

Danger

86609.550EA 0 EUR
86609.550
Dearomatized hydrocarbons EXXSOL DSP 100/140
Dearomatized hydrocarbons
Density: 0,745 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 68920-06-9
EINECS: 272-912-9
REACH: 01-2119473851-33

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR