Would you like to log in again?
96-well, round well with V-bottom storage plate, Falcon®
Supplier: Corning Life Science
Falcon®
734-0391EA 385 EUR InStock
734-0391
96-well, round well with V-bottom storage plate, Falcon®
Microplates HTS Microplates
Falcon® 96-well PP microplate, non treated, V-bottom is convenient for compound storage.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty