Bottles, wide neck, with NS stopper
Supplier: VITLAB
VITL1017940EA 347 EUR
VITL1017940 VITL1018940
Bottles, wide neck, with NS stopper
Bottles Plastic Bottles
Made from opaque, pigmented PP, with NS stopper made of PP.
  • Good chemical resistance
  • Ideal for long-term storage of liquids
  • Autoclavable at 121 °C (2 bar) according to DIN EN 285
For light-sensitive substances. Wider mouth for simple filling, also ideal for powders and paste-like materials.