Beakers, low form
Supplier: VITLAB
VITL112197EA 23.8 EUR
VITL112197 VITL112597 VITL112897 VITL112397 VITL112797 VITL112697 VITL112497 VITL112297
Beakers, low form
Beakers
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +260 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.