Wash head for wash bottles
Supplier: VITLAB
VITL83301EA 42.4 EUR
VITL83301 VITL83302 VITL83300 VITL83304 VITL83303
Wash head for wash bottles
Closures Bottle Closures
Made of PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
Wash bottle screw caps and spray tube with drawn-out tip, Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.