Wash bottles, narrow neck
Supplier: Vitlab
VITL108792EA 0 EUR
VITL108792
Wash bottles, narrow neck
Bottles Wash Bottles
PFA, translucent, screw cap made from ETFE, spray tube from FEP.
  • Excellent chemical resistance
  • Temperature resistance –200 to +260 °C
  • Can be used for less critical applications for which pure PFA is not necessary
With recycled PFA content. Thus, reasonably priced and environmentally friendly. Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.