Alcohol lamps
Supplier: GLASWARENFABRIK KARL HECHT
HECH43031010EA 38.6 EUR
HECH43031010
Alcohol lamps
Heaters Spirit Lamps
Glass alcohol lamps without lateral tubing, complete with wick and brass wick holder.
  • With a ground glass stopper
  • Glass cap reduces evoporation of alcohol when not in use