Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium ≥99.99% (metals basis excluding Sr)
|
Warning
|
Synonyms:
Barium(II) 2,2,6,6-tetramethylheptane-3,5-dionate hydrate
, Barium 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
41340.06EA
276
EUR
41340.06
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium ≥99.99% (metals basis excluding Sr)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium
Formula:
C₂₂H₃₈BaO₄ MW: 503,85 g/mol Boiling Pt: 225 °C (0,3 mmHg) Melting Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058709 CAS Number: 17594-47-7 UN: 1564 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...