Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III) ≥99%

Supplier: Thermo Scientific

Warning

Synonyms: Pr(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium(III)

A12259.06EA 111 EUR
A12259.06 A12259.03
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III) ≥99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III)
Formula: C₃₃H₅₇O₆Pr
Melting Pt: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00000027
CAS Number: 15492-48-5
EINECS: 239-522-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR