Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')samarium(III) ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: Sm(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)samarium(III)

A18004.03EA 53.1 EUR
A18004.03
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')samarium(III) ≥99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')samarium(III)
Formula: C₃₃H₅₇O₆Sm
Melting Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00015711
CAS Number: 15492-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR