Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III) ≥99.9% (metals basis), recrystallized

Supplier: Alfa Aesar
Warning

Synonyms: Fe(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III)

41862.03EA 56.3 EUR
41862.03 41862.06 41862.14
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III) ≥99.9% (metals basis), recrystallized
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III)
Formula: C₃₃H₅₇FeO₆
Boiling Pt: 300 °C (dec.)
Melting Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00067465
CAS Number: 14876-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR