Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III) ≥99%

Supplier: Alfa Aesar
Warning

Synonyms: Ru(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ruthenium(III)

44006.04EA 637 EUR
44006.04 44006.02
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III) ≥99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III)
Formula: C₃₃H₅₇O₆Ru
Boiling Pt: 250 °C (dec.)
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00269841
CAS Number: 38625-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR