Thallium(I) acetylacetonate ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: Thallium(I) 2,4-pentanedionate

78113.03EA 51.9 EUR
78113.03 78113.06
Thallium(I) acetylacetonate ≥97%
Thallium (I) acetylacetonate
Formula: C₅H₇O₂Tl
MW: 303,491 g/mol
Melting Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00013506
CAS Number: 25955-51-5
EINECS: 238-081-1
UN: 1707
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR