Hafnium trifluoroacetylacetonate

Supplier: Thermo Scientific
Warning

Synonyms: Hafnium 1,1,1-trifluoro-2,4-pentanedionate

19275.09EA 226 EUR
19275.09 19275.04 19275.02
Hafnium trifluoroacetylacetonate
Hafnium trifluoroacetylacetonate
Formula: C₂₀H₁₆F₁₂O₈Hf
Melting Pt: 128…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00078027
CAS Number: 17475-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR