Bis(tetramethylcyclopentadienyl)chromium(II) ≥98%

Supplier: Alfa Aesar
Danger

Synonyms: Bis(tetramethylcyclopentadienyl)chromium

H27396.03EA 87.8 EUR
H27396.03 H27396.06
Bis(tetramethylcyclopentadienyl)chromium(II) ≥98%
Bis(tetramethylcyclopentadienyl)chromium (II)
Formula: C₁₈H₂₆Cr
Melting Pt: 106…111 °C
MDL Number: MFCD01862451
CAS Number: 82066-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR