Bis(tetramethylcyclopentadienyl)manganese(II) ≥98%

Supplier: Thermo Scientific

Synonyms: Bis(tetramethylcyclopentadienyl)manganese

H27027.06EA 351 EUR
H27027.06 H27027.03
Bis(tetramethylcyclopentadienyl)manganese(II) ≥98%
Bis(tetramethylcyclopentadienyl)manganese (II)
Formula: C₁₈H₂₆Mn
MW: 297,34 g/mol
Melting Pt: 98…112 °C
MDL Number: MFCD01862454
CAS Number: 101932-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR