Hexaamminenickel(II) bromide ≥99.999% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
44482.14EA
111
EUR
44482.14
Hexaamminenickel(II) bromide ≥99.999% (metals basis)
Hexaamminenickel (II) bromide
Formula:
H₁₈Br₂N₆Ni MW: 320,68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039935 CAS Number: 13601-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...