Hexaamminenickel(II) iodide ≥99.999% (metals basis)

Supplier: Alfa Aesar
Danger

44481.14EA 150 EUR
44481.14
Hexaamminenickel(II) iodide ≥99.999% (metals basis)
Hexaamminenickel (II) iodide
Formula: H₁₈I₂N₆Ni
Storage Temperature: Ambient
MDL Number: MFCD00058808
CAS Number: 13859-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR