2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%

H55812.MDEA 20.8 EUR
ALFAH55812.MD ALFAH55812.06 ALFAH55812.03
2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
Purity: 98%
CAS No.: 49805-30-3
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 110°(230°F)
Melting Point: 54-58°
Boiling Point: 102-106°/0.25mm
Beilstein Reference: 508342
MDL No.: MFCD00213364
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR