(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one

Synonyms: 2-azabicyclo[2.2.1]hept-5-en-3-one

MAYBBTBG00205.1EA 0 EUR
MAYBBTBG00205.1 CHMPBTBG00205.250
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Formula: C₆H₇NO
MDL Number: MFCD00213364
CAS Number: 49805-30-3
EINECS: 421-830-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR